siRNA Details
| Gene Symbol: IFT22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001130820.2 | NP_001124292.1 | 4 | 1178 | ||
| NM_001130821.2 | NP_001124293.1 | 5 | 1299 | |||
| NM_001130822.2 | NP_001124294.1 | 4 | 1191 | |||
| NM_001287525.1 | NP_001274454.1 | 6 | 1341 | |||
| NM_001287526.1 | NP_001274455.1 | 3 | 1101 | |||
| NM_022777.3 | NP_073614.1 | 5 | 1268 | |||
| GenBank mRNA | AK023287.1 | - | 6 | 1299 | ||
| AK024179.1 | - | 5 | 1194 | |||
| AL157469.1 | - | 2 | 880 | |||

IFT22
Genomic Map