siRNA Details
| Gene Symbol: GAL3ST4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024637.4 | NP_078913.3 | 4 | 1695 | ||
| GenBank mRNA | AF289575.1 | - | 3 | 1490 | ||
| AF316113.1 | - | 4 | 1538 | |||
| AK022178.1 | - | 3 | 1334 | |||
| AK225890.1 | - | 3 | 1471 | |||
| AK301591.1 | - | 2 | 1133 | |||
| AL833824.1 | - | 3 | 1340 | |||
| BC012976.2 | - | 4 | 1514 | 3636329 | ||

GAL3ST4
Genomic Map