siRNA Details
| Gene Symbol: LRRK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024652.4 | NP_078928.3 | 21 | 3405 | ||
| XM_005254979.3 | XP_005255036.1 | 7 | 1063 | |||
| XM_011522012.2 | XP_011520314.1 | 23 | 3679 | |||
| XM_011522013.2 | XP_011520315.1 | 21 | 3388 | |||
| XM_011522014.2 | XP_011520316.1 | 21 | 3603 | |||
| XM_011522015.2 | XP_011520317.1 | 16 | 4198 | |||
| XM_011522016.2 | XP_011520318.1 | 19 | 2880 | |||
| XM_011522017.2 | XP_011520319.1 | 7 | 1070 | |||
| XM_011522018.2 | XP_011520320.1 | 7 | 1065 | |||
| XM_011522019.2 | XP_011520321.1 | 21 | 3603 | |||
| XM_017022570.1 | XP_016878059.1 | 11 | 1652 | |||
| GenBank mRNA | AB058693.1 | - | 7 | 1110 | ||
| AK131075.1 | - | 2 | 338 | |||
| BC144705.1 | - | 20 | 2957 | |||
| BC146769.1 | - | 7 | 1110 | |||
| BC171802.1 | - | 20 | 3079 | |||
| DQ013130.1 | - | 20 | 3043 | |||

LRRK1
Genomic Map