siRNA Details
| Gene Symbol: PTCD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001284403.1 | NP_001271332.1 | 7 | 779 | ||
| NM_001284404.1 | NP_001271333.1 | 8 | 909 | |||
| NM_001284405.1 | NP_001271334.1 | 9 | 1027 | |||
| NM_024754.4 | NP_079030.3 | 10 | 1106 | |||
| XM_005248601.2 | XP_005248658.1 | 9 | 986 | |||
| XM_005248603.2 | XP_005248660.1 | 9 | 962 | |||
| XM_017009866.1 | XP_016865355.1 | 9 | 967 | |||
| XM_017009867.1 | XP_016865356.1 | 8 | 879 | |||
| XM_017009868.1 | XP_016865357.1 | 8 | 848 | |||
| GenBank mRNA | AK022660.1 | - | 8 | 868 | ||
| AK056761.1 | - | 10 | 1770 | |||
| AK298750.1 | - | 7 | 770 | |||
| AK303639.1 | - | 8 | 878 | |||
| BC018720.1 | - | 10 | 1082 | 4552964 | ||
| CR457290.1 | - | 6 | 634 | |||

PTCD2
Genomic Map