siRNA Details
| Gene Symbol: MMRN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024756.2 | NP_079032.2 | 6 | 1623 | ||
| XM_005270153.1 | XP_005270210.1 | 6 | 1662 | |||
| XM_006717970.3 | XP_006718033.1 | 6 | 3602 | |||
| GenBank mRNA | AK023527.1 | - | 6 | 1570 | ||
| AK293822.1 | - | 5 | 904 | |||
| AK304727.1 | - | 6 | 1480 | |||
| AK308401.1 | - | 5 | 1464 | |||
| BC064415.1 | - | 6 | 1578 | |||
| BC072456.1 | - | 6 | 1519 | |||
| BC094744.1 | - | 6 | 1570 | 30343159 | ||

MMRN2
Genomic Map