siRNA Details
| Gene Symbol: TREML2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024807.3 | NP_079083.2 | 3 | 698 | ||
| XM_011514917.2 | XP_011513219.1 | 2 | 423 | |||
| GenBank mRNA | AF534824.1 | - | 3 | 697 | ||
| AK023755.1 | - | 3 | 697 | |||
| AY358171.1 | - | 3 | 699 | |||
| BC125078.1 | - | 3 | 697 | 40117710 | ||
| BC125079.1 | - | 3 | 697 | 40117711 | ||

TREML2
Genomic Map