siRNA Details
| Gene Symbol: TREML2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024807.3 | NP_079083.2 | 5 | 2639 | ||
| XM_011514917.2 | XP_011513219.1 | 4 | 2364 | |||
| GenBank mRNA | AJ420509.1 | - | 1 | 454 | ||
| AK023755.1 | - | 5 | 2640 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TREML2| Gene Symbol: TREML2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024807.3 | NP_079083.2 | 5 | 2639 | ||
| XM_011514917.2 | XP_011513219.1 | 4 | 2364 | |||
| GenBank mRNA | AJ420509.1 | - | 1 | 454 | ||
| AK023755.1 | - | 5 | 2640 | |||