siRNA Details
| Gene Symbol: GGNBP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024835.4 | NP_079111.1 | 5, 6 | 837 | ||
| XM_005257689.2 | XP_005257746.1 | 5, 6 | 813 | |||
| XM_011525265.2 | XP_011523567.1 | 4, 5 | 576 | |||
| XM_017025107.1 | XP_016880596.1 | 4, 5 | 550 | |||
| GenBank mRNA | AF126964.1 | - | 4, 5 | 568 | ||
| AF268387.1 | - | 5, 6 | 757 | |||
| AK024883.1 | - | 5, 6 | 793 | |||
| AK290687.1 | - | 4, 5 | 593 | |||
| AK292161.1 | - | 5, 6 | 806 | |||
| AK301614.1 | - | 2, 3 | 125 | |||
| BC137491.1 | - | 5, 6 | 703 | 9021491 | ||

GGNBP2
Genomic Map