siRNA Details
| Gene Symbol: SCO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004589.3 | NP_004580.1 | 2 | 385 | ||
| XM_005256751.3 | XP_005256808.1 | 2 | 352 | |||
| GenBank mRNA | AF026852.1 | - | 2 | 337 | ||
| AF183424.1 | - | 2 | 337 | |||
| AK074588.1 | - | 2 | 385 | |||
| AK098826.1 | - | 2 | 345 | |||
| AK315595.1 | - | 2 | 363 | |||
| BC015504.2 | - | 2 | 340 | 3914068 | ||
| DA571370.1 | - | 2 | 385 | |||

SCO1
Genomic Map