siRNA Details
| Gene Symbol: ZDHHC18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032283.2 | NP_115659.1 | 8 | 2191 | ||
| GenBank mRNA | AK056427.1 | - | 8 | 1943 | ||
| AL512765.1 | - | 8 | 1796 | |||
| BC066776.1 | - | 8 | 2191 | 30342061 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZDHHC18| Gene Symbol: ZDHHC18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032283.2 | NP_115659.1 | 8 | 2191 | ||
| GenBank mRNA | AK056427.1 | - | 8 | 1943 | ||
| AL512765.1 | - | 8 | 1796 | |||
| BC066776.1 | - | 8 | 2191 | 30342061 | ||