siRNA Details
| Gene Symbol: HSDL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001195822.1 | NP_001182751.1 | 8 | 1075 | ||
| NM_032303.4 | NP_115679.2 | 10 | 1294 | |||
| NR_036651.1 | 9 | 1130 | ||||
| XM_017015203.1 | XP_016870692.1 | 7 | 1528 | |||
| XM_017015204.1 | XP_016870693.1 | 9 | 1807 | |||
| XM_017015205.1 | XP_016870694.1 | 8 | 1643 | |||
| GenBank mRNA | AK090940.1 | - | 2 | 90 | ||
| AK098721.1 | - | 10 | 1265 | |||
| AK225946.1 | - | 7 | 827 | |||
| AK289929.1 | - | 10 | 1122 | |||
| AK292486.1 | - | 8 | 882 | |||
| AK301469.1 | - | 8 | 888 | |||
| AK303645.1 | - | 9 | 936 | |||
| AK313605.1 | - | 8 | 882 | |||
| AL833735.1 | - | 8 | 808 | |||
| AY093428.1 | - | 10 | 1133 | |||
| BC004331.1 | - | 8 | 904 | 3630835 | ||
| BC095451.1 | - | 10 | 1071 | 30405073 | ||

HSDL2
Genomic Map