siRNA Details
| Gene Symbol: FAM83A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001288587.2 | NP_001275516.1 | 2 | 1046 | ||
| NM_001321630.1 | NP_001308559.1 | 3 | 1214 | |||
| NM_032899.6 | NP_116288.2 | 4 | 1638 | |||
| NM_207006.2 | NP_996889.1 | 4 | 2723 | |||
| XM_005251087.2 | XP_005251144.1 | 2 | 843 | |||
| XM_011517338.1 | XP_011515640.1 | 3 | 1011 | |||
| GenBank mRNA | AK095301.1 | - | 4 | 2723 | ||
| BC052300.1 | - | 3 | 1027 | 6421272 | ||
| DQ280323.1 | - | 4 | 1611 | |||

FAM83A
Genomic Map