siRNA Details
Gene Symbol: SCRT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033129.3 | NP_149120.1 | 2 | 2868 | ||
GenBank mRNA | AY129025.1 | - | 2 | 2874 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: SCRT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033129.3 | NP_149120.1 | 2 | 2868 | ||
GenBank mRNA | AY129025.1 | - | 2 | 2874 |