siRNA Details
| Gene Symbol: FCHSD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033449.2 | NP_258260.1 | 20 | 2227 | ||
| XM_005268524.4 | XP_005268581.1 | 20 | 2254 | |||
| XM_006714803.3 | XP_006714866.1 | 19 | 2131 | |||
| XM_011537698.2 | XP_011536000.1 | 19 | 2172 | |||
| GenBank mRNA | AF086518.1 | - | 1 | 19 | ||
| AK000007.1 | - | 20 | 2211 | |||
| AK027281.1 | - | 1 | 218 | |||
| AK126203.1 | - | 2 | 1089 | |||
| AY217346.1 | - | 20 | 2227 | |||
| AY358478.1 | - | 11 | 1845 | |||
| BC047016.1 | - | 19 | 2169 | 5215830 | ||

FCHSD1
Genomic Map