siRNA Details
| Gene Symbol: TSPYL5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033512.2 | NP_277047.2 | 1 | 928 | ||
| GenBank mRNA | AB051537.1 | - | 1 | 867 | ||
| AK091445.1 | - | 1 | 944 | |||
| BC045630.1 | - | 1 | 928 | 5273220 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TSPYL5| Gene Symbol: TSPYL5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033512.2 | NP_277047.2 | 1 | 928 | ||
| GenBank mRNA | AB051537.1 | - | 1 | 867 | ||
| AK091445.1 | - | 1 | 944 | |||
| BC045630.1 | - | 1 | 928 | 5273220 | ||