siRNA Details
| Gene Symbol: WDR34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_052844.3 | NP_443076.2 | 4 | 701 | ||
| XM_011519179.2 | XP_011517481.1 | 4 | 635 | |||
| GenBank mRNA | AK302098.1 | - | 3 | 331 | ||
| AL555205.3 | - | 4 | 508 | |||
| BC001614.1 | - | 4 | 536 | |||
| BC011874.2 | - | 4 | 638 | 3161070 | ||
| BG177634.1 | - | 4 | 545 | |||

WDR34
Genomic Map