siRNA Details
| Gene Symbol: EMILIN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_052846.1 | NP_443078.1 | 4 | 1996 | ||
| GenBank mRNA | AB089149.1 | - | 4 | 1996 | ||
| AL137580.1 | - | 1 | 864 | |||
| BC101043.1 | - | 4 | 1772 | 40007928 | ||
| BC101044.2 | - | 4 | 1772 | |||
| BC101045.1 | - | 4 | 1772 | 40007932 | ||
| BC101046.1 | - | 4 | 1206 | |||

EMILIN3
Genomic Map