siRNA Details
| Gene Symbol: PLEKHG4B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_052909.3 | NP_443141.3 | 12 | 2605 | ||
| XM_006714446.3 | XP_006714509.1 | 14 | 3743 | |||
| XM_011513968.2 | XP_011512270.1 | 14 | 3740 | |||
| XM_011513969.2 | XP_011512271.1 | 14 | 3620 | |||
| XM_011513970.1 | XP_011512272.1 | 14 | 3332 | |||
| XM_011513971.2 | XP_011512273.1 | 14 | 3688 | |||
| XM_017009112.1 | XP_016864601.1 | 14 | 3744 | |||
| GenBank mRNA | AB067496.2 | - | 12 | 2605 | ||
| AK125163.1 | - | 1 | 434 | |||

PLEKHG4B
Genomic Map