siRNA Details
| Gene Symbol: CLNK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_052964.2 | NP_443196.2 | 16 | 985 | ||
| XM_011513775.2 | XP_011512077.1 | 16 | 1216 | |||
| XM_017007684.1 | XP_016863173.1 | 16 | 1216 | |||
| XM_017007685.1 | XP_016863174.1 | 16 | 1216 | |||
| GenBank mRNA | AB032369.1 | - | 14 | 794 | ||
| AB110420.1 | - | 16 | 1088 | |||
| AK093001.1 | - | 16 | 987 | |||
| BC137553.1 | - | 16 | 914 | 9021553 | ||
| BC137556.1 | - | 16 | 914 | 9021556 | ||

CLNK
Genomic Map