siRNA Details
| Gene Symbol: BPIFA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001319164.1 | NP_001306093.1 | 2 | 163 | ||
| NM_080574.3 | NP_542141.1 | 2 | 245 | |||
| GenBank mRNA | AF432917.1 | - | 1 | 97 | ||
| AK291854.1 | - | 2 | 245 | |||
| AY359055.1 | - | 2 | 163 | |||
| BC065726.1 | - | 2 | 172 | 30326122 | ||

BPIFA2
Genomic Map