siRNA Details
| Gene Symbol: TSPAN17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001006616.2 | NP_001006617.2 | 4 | 591 | ||
| NM_012171.2 | NP_036303.1 | 4 | 591 | |||
| NM_130465.4 | NP_569732.2 | 4 | 591 | |||
| XM_005265876.3 | XP_005265933.1 | 2 | 373 | |||
| GenBank mRNA | AK024427.1 | - | 4 | 481 | ||
| AY037146.1 | - | 2 | 313 | |||
| BC010405.2 | - | 4 | 413 | 3621871 | ||
| BC067105.1 | - | 4 | 476 | 6646576 | ||

TSPAN17
Genomic Map