siRNA Details
| Gene Symbol: SYCE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001143763.1 | NP_001137235.1 | 7, 8 | 577 | ||
| NM_001143764.1 | NP_001137236.1 | 7, 8 | 577 | |||
| NM_130784.2 | NP_570140.1 | 7, 8 | 500 | |||
| GenBank mRNA | AK314978.1 | - | 7, 8 | 527 | ||
| AY027807.1 | - | 7, 8 | 500 | |||
| AY027808.1 | - | 4, 5 | 1436 | |||
| BC034821.1 | - | 7, 8 | 527 | 5164386 | ||

SYCE1
Genomic Map