siRNA Details
| Gene Symbol: TMEM132D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133448.2 | NP_597705.2 | 9 | 4355 | ||
| GenBank mRNA | AB061814.1 | - | 9 | 4369 | ||
| AB075824.1 | - | 4 | 3882 | |||
| AK094656.1 | - | 1 | 1202 | |||
| AL833234.1 | - | 1 | 1478 | |||

TMEM132D
Genomic Map