siRNA Details
| Gene Symbol: PWWP2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138499.3 | NP_612508.3 | 2 | 1794 | ||
| XM_006717659.3 | XP_006717722.1 | 2 | 1796 | |||
| GenBank mRNA | AF318365.1 | - | 1 | 1445 | ||
| AK128663.1 | - | 1 | 1630 | |||
| BC011630.2 | - | 1 | 1266 | |||
| BC068574.1 | - | 2 | 1792 | 30344364 | ||

PWWP2B
Genomic Map