siRNA Details
| Gene Symbol: R3HDM4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138774.3 | NP_620129.2 | 8 | 811 | ||
| XM_011528416.2 | XP_011526718.1 | 8 | 815 | |||
| GenBank mRNA | BC012775.2 | - | 8 | 786 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

R3HDM4| Gene Symbol: R3HDM4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138774.3 | NP_620129.2 | 8 | 811 | ||
| XM_011528416.2 | XP_011526718.1 | 8 | 815 | |||
| GenBank mRNA | BC012775.2 | - | 8 | 786 | ||