siRNA Details
| Gene Symbol: BTBD16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001318189.1 | NP_001305118.1 | 8 | 901 | ||
| NM_144587.3 | NP_653188.2 | 8 | 898 | |||
| XM_011539239.2 | XP_011537541.1 | 8 | 1452 | |||
| XM_011539240.2 | XP_011537542.1 | 7 | 1307 | |||
| XM_017015636.1 | XP_016871125.1 | 7 | 1981 | |||
| XM_017015637.1 | XP_016871126.1 | 4 | 1606 | |||
| GenBank mRNA | AK058088.1 | - | 8 | 888 | ||
| BC109261.1 | - | 8 | 872 | 40025404 | ||
| BC109262.1 | - | 8 | 872 | 40025414 | ||

BTBD16
Genomic Map