siRNA Details
| Gene Symbol: ZNRF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147128.3 | NP_667339.1 | 4 | 1773 | ||
| GenBank mRNA | AF513707.1 | - | 4 | 1773 | ||
| AF527533.1 | - | 4 | 1100 | |||
| AK025240.1 | - | 3 | 215 | |||
| AY054122.1 | - | 5 | 537 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNRF2| Gene Symbol: ZNRF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147128.3 | NP_667339.1 | 4 | 1773 | ||
| GenBank mRNA | AF513707.1 | - | 4 | 1773 | ||
| AF527533.1 | - | 4 | 1100 | |||
| AK025240.1 | - | 3 | 215 | |||
| AY054122.1 | - | 5 | 537 | |||