siRNA Details
| Gene Symbol: DEPDC4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001319311.1 | NP_001306240.1 | 11 | 1605 | ||
| NR_135073.1 | 8 | 1260 | ||||
| XM_017018781.1 | XP_016874270.1 | 10 | 1566 | |||
| GenBank mRNA | AK090824.1 | - | 8 | 1260 | ||

DEPDC4
Genomic Map