siRNA Details
| Gene Symbol: SLC35G1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134658.1 | NP_001128130.1 | 3 | 1205 | ||
| NM_153226.2 | NP_694958.1 | 3 | 1202 | |||
| XM_011539371.2 | XP_011537673.1 | 3 | 1170 | |||
| GenBank mRNA | AK091309.1 | - | 3 | 1202 | ||
| BC041432.1 | - | 2 | 934 | |||
| BC104814.1 | - | 3 | 1176 | 8143817 | ||
| BC143283.1 | - | 3 | 1125 | 9051786 | ||
| BC143284.1 | - | 3 | 1176 | 9051787 | ||

SLC35G1
Genomic Map