siRNA Details
| Gene Symbol: PSTK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153336.2 | NP_699167.2 | 7 | 1614 | ||
| XM_017015640.1 | XP_016871129.1 | 6 | 1069 | |||
| GenBank mRNA | AK094271.1 | - | 6 | 1719 | ||
| AK127173.1 | - | 5 | 2104 | |||
| BC035344.1 | - | 7 | 1614 | 5185692 | ||

PSTK
Genomic Map