siRNA Details
| Gene Symbol: RNF19B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127361.2 | NP_001120833.1 | 5 | 1346 | ||
| NM_001300826.1 | NP_001287755.1 | 5 | 1346 | |||
| NM_153341.3 | NP_699172.2 | 5 | 1349 | |||
| XM_006710356.2 | XP_006710419.1 | 5 | 1802 | |||
| XM_006710357.3 | XP_006710420.1 | 5 | 1802 | |||
| XM_006710358.3 | XP_006710421.1 | 5 | 1802 | |||
| GenBank mRNA | AK074486.1 | - | 5 | 649 | ||
| AK131439.1 | - | 6 | 1043 | |||
| BC020595.2 | - | 1 | 54 | |||
| BC062374.1 | - | 5 | 823 | |||
| BC113560.1 | - | 5 | 716 | 8322612 | ||
| BC143687.1 | - | 5 | 713 | 9052201 | ||

RNF19B
Genomic Map