siRNA Details
| Gene Symbol: PLEKHH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172069.3 | NP_742066.2 | 8 | 1345 | ||
| XM_017003351.1 | XP_016858840.1 | 7 | 1332 | |||
| XM_017003352.1 | XP_016858841.1 | 8 | 1345 | |||
| XM_017003353.1 | XP_016858842.1 | 8 | 1345 | |||
| GenBank mRNA | AB095948.1 | - | 6 | 1131 | ||
| AK124381.1 | - | 1 | 92 | |||
| AK126308.1 | - | 8 | 1342 | |||
| AL832207.1 | - | 8 | 1338 | |||
| AL833546.1 | - | 9 | 1540 | |||
| BC063310.1 | - | 8 | 1372 | 30342490 | ||

PLEKHH2
Genomic Map