siRNA Details
| Gene Symbol: PLEKHH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172069.3 | NP_742066.2 | 30 | 6700 | ||
| XM_017003351.1 | XP_016858840.1 | 29 | 6687 | |||
| XM_017003353.1 | XP_016858842.1 | 31 | 6811 | |||
| GenBank mRNA | AB095948.1 | - | 28 | 6486 | ||
| AL832207.1 | - | 30 | 6693 | |||
| AL833473.1 | - | 5 | 2836 | |||

PLEKHH2
Genomic Map