siRNA Details
| Gene Symbol: C2orf73 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100396.1 | NP_001093866.1 | 5 | 740 | ||
| XM_005264133.1 | XP_005264190.1 | 4 | 532 | |||
| XM_005264134.3 | XP_005264191.1 | 4 | 615 | |||
| XM_011532510.2 | XP_011530812.1 | 5 | 810 | |||
| XM_011532513.2 | XP_011530815.1 | 4 | 713 | |||
| XM_017003323.1 | XP_016858812.1 | 5 | 635 | |||
| XM_017003324.1 | XP_016858813.1 | 4 | 698 | |||
| GenBank mRNA | AK097617.1 | - | 4 | 532 | ||
| BC031669.1 | - | 5 | 703 | |||
| BC126244.1 | - | 4 | 501 | 8991960 | ||
| BC126246.1 | - | 4 | 501 | 8991962 | ||
| BC144191.1 | - | 3 | 385 | |||
| BC144192.1 | - | 5 | 535 | 9052712 | ||

C2orf73
Genomic Map