siRNA Details
| Gene Symbol: ZDHHC23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173570.4 | NP_775841.2 | 5, 6 | 1523 | ||
| XM_005247269.1 | XP_005247326.1 | 5, 6 | 1467 | |||
| XM_017006086.1 | XP_016861575.1 | 5, 6 | 1458 | |||
| GenBank mRNA | AK127996.1 | - | 6, 7 | 1468 | ||
| BC035230.1 | - | 5, 6 | 1523 | 4824088 | ||

ZDHHC23
Genomic Map