siRNA Details
| Gene Symbol: ZDHHC23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001320466.1 | NP_001307395.1 | 5 | 4986 | ||
| NM_001320467.1 | NP_001307396.1 | 5 | 4878 | |||
| NM_001320468.1 | NP_001307397.1 | 5 | 4977 | |||
| NM_173570.4 | NP_775841.2 | 6 | 2697 | |||
| NR_135271.1 | 7 | 2553 | ||||
| XM_005247269.1 | XP_005247326.1 | 6 | 2641 | |||
| XM_005247270.3 | XP_005247327.1 | 4 | 4117 | |||
| XM_011512618.2 | XP_011510920.1 | 5 | 4826 | |||
| XM_011512619.1 | XP_011510921.1 | 6 | 3840 | |||
| XM_017006084.1 | XP_016861573.1 | 5 | 4793 | |||
| XM_017006085.1 | XP_016861574.1 | 6 | 3819 | |||
| XM_017006086.1 | XP_016861575.1 | 6 | 2632 | |||
| XM_017006087.1 | XP_016861576.1 | 6 | 3919 | |||
| GenBank mRNA | AF131818.1 | - | 1 | 1413 | ||
| AK022842.1 | - | 1 | 1814 | |||
| AK127260.1 | - | 2 | 3819 | |||
| BC035230.1 | - | 6 | 2697 | 4824088 | ||

ZDHHC23
Genomic Map