siRNA Details
| Gene Symbol: ARL10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173664.5 | NP_775935.1 | 4 | 1335 | ||
| XM_011534531.2 | XP_011532833.1 | 4 | 829 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ARL10| Gene Symbol: ARL10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173664.5 | NP_775935.1 | 4 | 1335 | ||
| XM_011534531.2 | XP_011532833.1 | 4 | 829 | |||