siRNA Details
| Gene Symbol: SPATA19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001291992.1 | NP_001278921.1 | 1, 2 | 171 | ||
| NM_174927.2 | NP_777587.1 | 1, 2 | 171 | |||
| GenBank mRNA | AF521713.1 | - | 1, 2 | 107 | ||
| AK098717.1 | - | 1, 2 | 130 | |||
| BC058039.1 | - | 1, 2 | 138 | 6619434 | ||
| BX096445.1 | - | 1, 2 | 171 | |||

SPATA19
Genomic Map