siRNA Details
| Gene Symbol: TAAR9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175057.3 | NP_778227.3 | 1 | 1005 | ||
| GenBank mRNA | AY183469.1 | - | 1 | 1003 | ||
| BC069083.1 | - | 1 | 1005 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TAAR9| Gene Symbol: TAAR9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175057.3 | NP_778227.3 | 1 | 1005 | ||
| GenBank mRNA | AY183469.1 | - | 1 | 1003 | ||
| BC069083.1 | - | 1 | 1005 | |||
Set Membership: |
|