siRNA Details
| Gene Symbol: ZNF598 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178167.3 | NP_835461.2 | 8 | 1296 | ||
| GenBank mRNA | AK024487.1 | - | 6 | 2021 | ||
| AL834428.1 | - | 2 | 98 | |||
| BC010990.2 | - | 4 | 509 | |||
| BC041015.1 | - | 2 | 81 | |||
| BC050477.1 | - | 8 | 1237 | 6055142 | ||

ZNF598
Genomic Map