siRNA Details
| Gene Symbol: AGBL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178563.3 | NP_848658.3 | 7 | 1530 | ||
| XM_011516122.2 | XP_011514424.1 | 6 | 1336 | |||
| XM_017012136.1 | XP_016867625.1 | 7 | 2852 | |||
| XM_017012137.1 | XP_016867626.1 | 7 | 3134 | |||
| XM_017012138.1 | XP_016867627.1 | 6 | 1336 | |||
| XM_017012139.1 | XP_016867628.1 | 6 | 1336 | |||
| XM_017012140.1 | XP_016867629.1 | 6 | 1336 | |||
| XM_017012141.1 | XP_016867630.1 | 6 | 2669 | |||
| XM_017012142.1 | XP_016867631.1 | 6 | 1336 | |||
| XM_017012143.1 | XP_016867632.1 | 6 | 1336 | |||
| GenBank mRNA | AK302827.1 | - | 7 | 1530 | ||
| BC030651.2 | - | 7 | 1505 | 5267615 | ||

AGBL3
Genomic Map