siRNA Details
| Gene Symbol: SKA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100595.1 | NP_001094065.1 | 3 | 586 | ||
| NM_182620.3 | NP_872426.1 | 4 | 588 | |||
| XM_011524745.2 | XP_011523047.1 | 4 | 422 | |||
| GenBank mRNA | AK022820.1 | - | 3 | 143 | ||
| AK056473.1 | - | 4 | 340 | |||
| BC017873.1 | - | 4 | 349 | 4685972 | ||
| BC106003.1 | - | 4 | 313 | 6143473 | ||
| DA133216.1 | - | 3 | 324 | |||
| DB158057.1 | - | 2 | 324 | |||

SKA2
Genomic Map