siRNA Details
| Gene Symbol: CLEC4G | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001244856.1 | NP_001231785.1 | 9 | 1115 | ||
| NM_198492.3 | NP_940894.1 | 9 | 1151 | |||
| GenBank mRNA | AY203939.1 | - | 9 | 1073 | ||
| AY358431.1 | - | 10 | 1149 | |||
| AY423706.1 | - | 9 | 1151 | |||
| BC039679.1 | - | 5 | 738 | |||

CLEC4G
Genomic Map