siRNA Details
| Gene Symbol: ZNF710 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198526.3 | NP_940928.2 | 5 | 2306 | ||
| GenBank mRNA | AK094712.1 | - | 5 | 2277 | ||
| AK160373.1 | - | 4 | 996 | |||
| AL831826.1 | - | 4 | 777 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF710| Gene Symbol: ZNF710 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198526.3 | NP_940928.2 | 5 | 2306 | ||
| GenBank mRNA | AK094712.1 | - | 5 | 2277 | ||
| AK160373.1 | - | 4 | 996 | |||
| AL831826.1 | - | 4 | 777 | |||