siRNA Details
| Gene Symbol: SP6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001258248.1 | NP_001245177.1 | 2 | 3600 | ||
| NM_199262.2 | NP_954871.1 | 2 | 3589 | |||
| XM_006722115.3 | XP_006722178.1 | 2 | 3552 | |||
| GenBank mRNA | AK127707.1 | - | 2 | 3641 | ||
| AK127850.1 | - | 2 | 3589 | |||

SP6
Genomic Map