siRNA Details
| Gene Symbol: SLC16A13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_201566.2 | NP_963860.1 | 4 | 1531 | ||
| GenBank mRNA | BC109286.1 | - | 4 | 1219 | ||
| BC131739.1 | - | 4 | 1219 | |||
| BC142618.1 | - | 4 | 1531 | 40147405 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SLC16A13| Gene Symbol: SLC16A13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_201566.2 | NP_963860.1 | 4 | 1531 | ||
| GenBank mRNA | BC109286.1 | - | 4 | 1219 | ||
| BC131739.1 | - | 4 | 1219 | |||
| BC142618.1 | - | 4 | 1531 | 40147405 | ||