siRNA Details
| Gene Symbol: C9orf131 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_203299.3 | NP_976044.2 | 1 | 173 | ||
| GenBank mRNA | AK097595.1 | - | 1 | 139 | ||
| AK302123.1 | - | 1 | 115 | |||
| BC045643.1 | - | 1 | 152 | 5298387 | ||
| DB068600.1 | - | 1 | 173 | |||

C9orf131
Genomic Map