siRNA Details
| Gene Symbol: TMEM81 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_203376.1 | NP_976310.1 | 1 | 1167 | ||
| GenBank mRNA | AK303432.1 | - | 1 | 1224 | ||
| AY359081.1 | - | 1 | 1167 | |||
| BC061592.1 | - | 1 | 1110 | 6202253 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TMEM81| Gene Symbol: TMEM81 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_203376.1 | NP_976310.1 | 1 | 1167 | ||
| GenBank mRNA | AK303432.1 | - | 1 | 1224 | ||
| AY359081.1 | - | 1 | 1167 | |||
| BC061592.1 | - | 1 | 1110 | 6202253 | ||