siRNA Details
| Gene Symbol: VNN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004666.2 | NP_004657.2 | 1 | 169 | ||
| GenBank mRNA | AJ132099.1 | - | 1 | 163 | ||
| AK290425.1 | - | 1 | 166 | |||
| AK314265.1 | - | 1 | 166 | |||
| BC096265.3 | - | 1 | 163 | 40006204 | ||
| BC096266.3 | - | 1 | 163 | 40006205 | ||
| BC096267.3 | - | 1 | 163 | 40006207 | ||
| BC096268.1 | - | 1 | 162 | 40006208 | ||
| BJ995136.1 | - | 1 | 169 | |||
| U39664.1 | - | 1 | 164 | |||

VNN1
Genomic Map