siRNA Details
| Gene Symbol: IER5L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_203434.2 | NP_982258.2 | 1 | 1563 | ||
| GenBank mRNA | BC064028.1 | - | 2 | 1194 | 6526871 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

IER5L| Gene Symbol: IER5L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_203434.2 | NP_982258.2 | 1 | 1563 | ||
| GenBank mRNA | BC064028.1 | - | 2 | 1194 | 6526871 | |